Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
GlenGuard® Work Pants, VF Imagewear
Supplier: VF Imagewear
4295NB-34-32 4295NB-44-34 4295NB-46-36 4295NB-34-34 4295NB-30-28 4295NB-36-34 4295NB-36-32 4295NB-46-28 4295NB-28-32 4295NB-40-28 4295NB-40-32 4295NB-44-36 4295NB-38-32 4295NB-36-36 4295NB-42-28 4295NB-38-30 4295NB-36-28 4295NB-44-32 4295NB-32-30 4295NB-28-30 4295NB-28-34 4295NB-42-34 4295NB-44-30 4295NB-40-30 4295NB-32-32 4295NB-46-34 4295NB-46-30 4295NB-40-36 4295NB-46-32 4295NB-36-30 4295NB-42-30 4295NB-30-30 4295NB-32-28 4295NB-42-36 4295NB-38-28 4295NB-30-32 4295NB-40-34 4295NB-38-36 4295NB-28-28 4295NB-30-34 4295NB-32-34 4295NB-42-32 4295NB-34-30 4295NB-38-34 4295NB-44-28 4295NB-34-28
76123-334EA 121.25 USD InStock
76123-334 76123-312 76123-354 76123-310 76123-332 76123-316 76123-338 76123-314 76123-336 76123-294 76123-096 76123-292 76123-094 76123-352 76123-330 76123-298 76123-350 76123-296 76123-098 76123-290 76123-092 76123-090 76123-308 76123-302 76123-346 76123-324 76123-288 76123-300 76123-344 76123-322 76123-328 76123-306 76123-304 76123-326 76123-348 76123-084 76123-282 76123-280 76123-088 76123-286 76123-342 76123-320 76123-086 76123-284 76123-340 76123-318
GlenGuard® Work Pants, VF Imagewear
Workwear
Made with solid brass nail button and zipper closure, GlenGuard® work pants also feature bartacks at stress points for durability.
  • Arc rating: 9.5
  • 5.3 oz. GlenGuard® material
Pockets include two front slash pockets and two rear welt pockets.
Certifications: US Standards: NFPA 2112, NFPA 70E, ASTM F1506
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty