Would you like to log in again?
Rebel Hybrid Microscope Bundle, Upright and Inverted, Discover Echo
Supplier: ECHO
Rebel
RCND2001 RBLT3000VWR RCND2002
76307-546EA 1215.02 USD InStock
76307-546 76307-544 76359-898
Rebel Hybrid Microscope Bundle, Upright and Inverted, Discover Echo
Microscopes
Available NOW, no lead time!!!! Bundle kit includes Rebel body, 4x Ph, 10x Ph, and 20x Ph LWD phase contrast objectives and LWD phase condenser kit.
  • ECHO Pro: powerful yet simple software reimagined to collaboratively view, capture, and share images
  • Ultra high resolution 13" touch display
  • 8MP CMOS color camera
  • Plan achromat and apochromat options available
  • XY mechanical stage option available
  • Ergonomic biosafety cabinet and adjustable height table options available
Rebel was designed from the ground up to improve microscopy.
Versatility: Hybrid microscope easily converts between UPRIGHT and INVERTED configurations. Brightfield and phase contrast modes available. Eyepiece option available as well.
Productivity: Our award-winning touch interface is incredibly easy to use. Eyepieces have been replaced with a 13” iPad Pro with Retina Display to view and capture stunning crystal-clear images.
Rebel True Ergo Table and Rebel Logic + Biosafety cabinet by Labconco bring the microscope stage down flush to the tabletop, setting a new precendent in microscope ergonomics.
Ordering information: Microscope includes a one-year warranty on electrical, and five-year warranty on mechanical parts and optics. Contact your VWR representative to demo the the newest innovations in microscopy.
Delivery information: The Bundle kit includes Rebel Body, 4x Ph, 10x Ph, and 20x Ph LWD Phase Contrast Objectives and LWD Phase Condenser Kit. Dual Focus Control Module, Intelligent Nosepiece, 13 inch iPad Pro with Locking Stand, 8mp CMOS Color Camera, Power Cord, Microscope Cover, and Lens Cloth. This kit is ideal for tissue culture.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty