Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Extech™ Multi-Field EMF Meter, Flir®

Supplier: Flir
EMF450
75853-972EA 532.67 USD InStock
75853-972
Extech™ Multi-Field EMF Meter, Flir®
Electromagnetic Field Meters
Convenient 3 in 1 meter, measure and display Magnetic Field, Electric Field and Radio Frequency Strength.

  • Multi-Field measurements: Magnetic Field, Electric Field, and Radio Frequency (RF) Strength
  • Magnetic Field Triple Axis Sensor (X, Y, Z)
  • Audible alarm
  • 2.4" (240×320 pixels) color TFT display
  • Adjustable screen brightness (low, medium, high)
  • Store up to 20 RF historical records
  • Data Hold and Auto Power Off with disable feature

The convenient 3-in-1 EMF meter which simultaneously measures and displays Magnetic Field, Electric Field and Radio Frequency Strength. Ergonomic pocket-sized design with an easy-to-navigate menu. Large vivid color TFT display shows digital readings, bargraphs, and trending graph. Magnetic Field, Electric Field, and Radio Frequency (RF) Strength measurements. Store up to 20 RF historical records. Data Hold and Auto Power Off with disable feature.

Certifications: CE Marked.

Delivery information: Complete with 3 AAA batteries and USB power supply cable.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty