Would you like to log in again?
Extech™ Coating Thickness Tester, Flir®
Supplier: Flir
CG104
75853-974EA 390.19 USD InStock
75853-974
Extech™ Coating Thickness Tester, Flir®
Coating Testers
Offers non-destructive measurements of the thickness of ferrous and non-ferrous substrates.
  • Dual technology provides automatic recognition of ferrous (iron and steel) and non-ferrous (copper, aluminum, zinc, bronze, brass, etc) substrates
  • Magnetic induction for ferrous substrates
  • Eddy current measurement for non-ferrous substrates
  • 180° reversible LCD display with LED backlight
  • Store/recall up to 255 measurements
  • Audible high/low limit alarms
  • Substrate zeroing and two-point calibration function
The CG104 is a pocket-sized coating thickness tester which offers quick one-handed operation and non-destructive measurements of the thickness of ferrous and non-ferrous substrates. Its unique reversible display allows you to take readings in challenging angles of area being measured, plus it has an easy-to use menu system and backlit display feature. Built-in memory stores/recalls up to 255 readings.
Certifications: CE Marked.
Delivery information: Complete with two AAA batteries, calibration standards (one ferrous plate, one non-ferrous plate and one standard coating plate film), wrist strap and soft case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty