Would you like to log in again?
Clarity FOB Controls, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-FOBCTLS
95064-014CS 4079.44 USD InStock
95064-014
Clarity FOB Controls, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Can be used to validate Clarity IFOBT and other brand rapid IFOB tests. Contains 1 vial of positive and 1 vial of negative control.
  • 1 ml/vial
  • Room temperature storage
  • 16-18 months dating at the time of purchase
Can be used as training tool.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty