Would you like to log in again?
Extech™ Portable Turbidity Meter, Flir®
Supplier: Extech
TB400
10111-920EA 1127.51 USD InStock
10111-920
Extech™ Portable Turbidity Meter, Flir®
Turbidity Meters
Portable design allows you to take measurement on-site instead of bringing solution back to the lab.
  • Large big-digit LCD and splash-proof front panel
  • Measures up to 1000 NTU (Nephelometric Turbidity Unit) with 0.01 NTU resolution
  • Microprocessor-based circuitry assures high accuracy and repeatable readings
  • Requires only a 10 ml sample size
  • 2-point calibration, Auto Power Off
  • Battery operated for field and on-site testing
Portable Turbidity Meter that tests water, food, beverages and other aqueous solutions. Meter comes complete with all the accessories to conveniently test the turbidity of municipal water, food and beverage water, and other aqueous solutions where fluid clarity is important. Portable design allows you to take measurement on-site instead of bringing solution back to the lab.
Certifications: CE Marked.
Delivery information: Complete with 0 NTU standard solution test bottle, 100 NTU standard solution test bottle, cleaning solution (distilled water), (2) test bottles and covers, cleaning cloth, six AAA batteries, and hard carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty