Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Extech™ Personal Noise Dosimeter with USB Interface, Flir®
Supplier: Flir
SL400
75870-668EA 1223.49 USD InStock
75870-668
Extech™ Personal Noise Dosimeter with USB Interface, Flir®
Sound Meters
Perform OSHA/IEC noise accumulation surveys to determine total sound exposure over an 8-hour period.
  • Measures frequency weighted noise exposure and peak (C, Z) sound level
  • Datalogs up to 999,999 readings when used as a sound level meter with sampling times between 1 second and 24 hours
  • Sound level mode: displays sound level, min/max, time-averaged sound level (Leq), peak, and sound exposure level (SEL)
  • Adjustable Criterion Level, Exchange Rate and Threshold
  • One user-defined measurement setup
  • Optional 94 dB calibrator to perform before and after calibration checks per ANSI standards
  • USB interface with Windows®-compatible software to control setup and retrieve stored events or analyze real-time measurements
The Personal Noise Dosimeter with USB Interface. For employees that are exposed to loud noise, sound level monitoring is essential to determine what protective measures are required. The compact, lightweight Dosimeter with USB interface designed to test noise exposure and provide noise surveys for compliance with OSHA, MSHA, DOD, ACGIH, and ISO standards.
Certifications: CE Marked.
Delivery information: Includes PC software to control setup and retrieve stored events or analyze real-time measurements. Complete with belt clip, clip-on 0.5" (12.7 mm) microphone with 32" (81.3 cm) cable and windscreen, 9 V battery, software, USB cable, and carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty