Would you like to log in again?
Servus® SF Overboots, Honeywell Safety
Supplier: Honeywell Safety
Servus™ SF
11001-BLM-1XL 11003-BLM-MED 11003-BLM-XSM
89370-804CS 566.82 USD InStock
89370-804 89370-798 89370-800
Servus® SF Overboots, Honeywell Safety
Shoes and Boots
PVC dual-compound overboots are formulated for long-lasting wear and durability. Supersized, SF overboots fit over large work and recreational footwear to protect them from liquids, dirt, mud and other deteriorating elements.
  • 100% waterproof protection to keep work boots, and feet, dry
  • Over-sized design permits fit over standard work boots
  • Protection from harsh environments increases the longevity of expensive footwear
  • Extra large molded kick-off lugs for easy, hands-free removal
  • Boot buttons are molded to eliminate seams, providing waterproof protection
  • Black upper, yellow outsole
Servus® SF overboots feature PVC injection molded construction. These black boots are available in a height of 4 or 12", with an extra-wide grip outsole and a stretchy top. The dual compounding molds the uppers to the slip-resistant outsoles. Tabbed closure loops facilitate one-hand closure even when wearing gloves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty