Would you like to log in again?
Ranger® Flex-Gard Heavy-Duty Rubber Boots, Honeywell Safety
Supplier: Honeywell Safety
Ranger® Flex-Gard
2169-ODM-140 2169-ODM-050 2169-ODM-070 2149/12 2149/8 2149/9 2169-ODM-090 2169-ODM-110 2149/13 2149/7 2169-ODM-060 2149/11 2169-ODM-080 2169-ODM-130 2149/6 2169-ODM-120 2169-ODM-100 2149/10
89370-726CS 1381.73 USD InStock
89370-726 89370-748 89370-736 89370-728 89370-738 89370-744 89370-722 89370-732 89370-754 89370-746 89370-724 89370-756 89370-734 89370-740 89370-750 89370-742 89370-752 89370-730
Ranger® Flex-Gard Heavy-Duty Rubber Boots, Honeywell Safety
Shoes and Boots
Premium grade rubber is formulated to ensure superior performance and long term use. Ranger® heavy-duty boots feature highly abrasion-resistant outsoles designed for wear resistance and traction in rough terrain.
  • 100% waterproof construction
  • Steel toe impact protection
  • Ozone-resistant rubber remains flexible in cold weather
  • Tough, comfortable fabric linings for easy on/off
  • Built-in cushion insole adds an extra comfort layer
  • Boots are available in 12 or 16" heights
All boots feature flex-gard metatarsal protection, SAFE-STEP puncture-resistant steel midsoles, and knurled abrasion-resistant overlays in high wear areas to increase the life span of the footwear. Steel toe impact protection is standard, along with a heavy TracTread™ outsole. The 12" boot features a 3-eyelet lace-up top, while the 16" boot is equipped with a single top strap for a self-adjusting and secure fit.
Certifications: Boots meet ASTM F2413-11 M I/75 C/75 Mt/75 PR.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty