Would you like to log in again?
KIMBLE® TLC Labeling Template, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
416450-0000
KT416450-0000EA 256.07 USD InStock
KT416450-0000
KIMBLE® TLC Labeling Template, DWK Life Sciences
TLC Plate Applicators
A transparent labeling template designed to fit over TLC plates without disturbing the absorbent layer.
  • Origin and 10 cm bold reference lines are provided to enable fast calculation of Rf-values
The apparatus features spotting guide ruler, compound and solvent travel distance markers, and circles for estimation of spot sizes.
L×W×H: 220×200×10 mm
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty