Would you like to log in again?
Clarity Cholesterol Meter, Clarity Diagnostics
Supplier: Clarity Diagnostics
Clarity
DTG-CHGLMETER DTG-CHSTRIPS
95064-026CS 3784.99 USD InStock
95064-026 95064-028
Clarity Cholesterol Meter, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Analysers
The Clarity total cholesterol meter is ideal for quantitative detection of total cholesterol in capillary whole blood samples.
  • Small blood drop - only requires 10 µl of whole blood
  • Results in 30 seconds
  • Highly accurate and reliable
  • 90 day open vial stability of Clarity cholesterol test strips
Ideally suited for use in physicians' offices and health fairs.
Certifications: FDA cleared and CLIA waived.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty