Would you like to log in again?
Avantor® Hichrom, Unions
Supplier: VWR
Avantor® Hichrom
HI-089 HI-495
76399-042EA 115.06 USD InStock
76399-042 76399-374
Avantor® Hichrom, Unions
Fittings and Connectors
These unions are made of PEEK.
  • Use to connect two pieces of capillary tubing, using tubing sleeves
  • True ZDV union
  • Includes fittings and gauge plug
  • Pressure rated to 4000 psi
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty