Would you like to log in again?
CS 21 Series Fixed Transmitters, Toxic and Combustible, Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GfG Instrumentation
2214122 2214131 2214013 2214130 2214032 2214028 2214129
76334-872EA 1689.32 USD InStock
76334-872 76334-960 76334-964 76334-874 76334-962 76334-966 76334-958
CS 21 Series Fixed Transmitters, Toxic and Combustible, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The CS 21 transmitter has a high sensitivity to a broad range of toxic and combustible gases.
  • High sensitivity to a broad range of toxic and combustible gases
  • Long sensor life
  • Fast response
Wall mount installation is straightforward. The distance between transmitter and controller can be greater than 500 meters. A rugged aluminum casing protects the sensor and electronics from splashes of water and dirt. The transmitter must not be operated in EX-areas. For measurement of combustible gases in the LEL range the gas needs to be sampled by pump. In this case the EX-protection needs to be guaranteed with suitable external components (e.g. flame‑protection).
The CS 21 transmitter uses the proven chemosorption detection principle for detection of toxic gases, vapors and combustibles. The rapid response of the transmitter provides opportunity to take counter measures and thereby limit damages. The long life of the sensors makes the CS 21 a low-cost solution for your detection needs. A transmitter can be continuously operated for more than 5 years.
Ordering information: For available accessories, calibration kits or additional information, contact your Avantor representative and see the attached datasheet and manual.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty