Would you like to log in again?
CS 21 Series Fixed Transmitters (Freons), Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GfG Instrumentation
2214017 2214021 2214020 2214036
76334-974EA 1805.81 USD InStock
76334-974 76334-972 76334-978 76334-976
CS 21 Series Fixed Transmitters (Freons), Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The CS 21 transmitter has a high sensitivity to a broad range of refrigerants.
  • High sensitivity to a broad range of refrigerants
  • Long sensor life
  • Fast response
The CS 21 transmitter uses the proven chemosorption detection principle for detection of toxic gases, vapors and combustibles. The rapid response of the transmitter provides opportunity to take counter measures and thereby limit damages. The long life of the sensors makes the CS 21 a low-cost solution for your detection needs. A transmitter can be continuously operated for more than 5 years.
Wall mount installation is straightforward. The distance between transmitter and controller can be greater than 500 meters. A rugged aluminum casing protects the sensor and electronics from splashes of water and dirt. The transmitter must not be operated in EX-areas. For measurement of combustible gases in the LEL range the gas needs to be sampled by pump. In this case the EX-protection needs to be guaranteed with suitable external components (e.g. flame‑protection).
Ordering information: For available sensors, calibration kits and accessories or more information, contact your Avantor representative and see the attached datasheet and manual.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty