Would you like to log in again?
Vernier Go Direct® Charge Station
Supplier: Vernier
GDX-CRG
470315-120EA 88.99 USD InStock
470315-120
Vernier Go Direct® Charge Station
Power Supplies
The Go Direct® charge station is the perfect solution for charging Go Direct sensors.
Each charge station has sixteen charging ports - eight USB and eight wand-style sensor ports. LED lights on the Go Direct sensors will show charging status. The Go Direct Charge Station is also compatible with Go Wireless devices.
Ordering information: USB cables not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty