Would you like to log in again?
Celestial Star Globe
Supplier: EISCO SCIENTIFIC LLC
BD0077CSG
470324-780EA 89 USD InStock
470324-780
Celestial Star Globe
Globes
Perfect for introductory-level astronomy students.
  • 12" Globe diameter
  • 17.5" Tall (overall model)
  • A 3" detailed earth globe is fitted inside the large star globe
This stunning star globe illustrates the effects of Earth’s rotation on its axis and its revolution around the sun in addition to depicting the constellations, Milky Way, and major stars in three dimensions. Color-coding and clear silk-screened names make it easy to identify the many stars, while celestial latitude and longitude scales provide orientation. A detailed 3" Earth globe is centered in the larger star globe. Both globes can be set to the positions of the stars and planets for any time and place on Earth in the past, present, or future.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty