Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
STAT-SKREEN DOA & POC Rapid Testing Controls, KOVA International
Supplier: Kova International
PREGNANCY-SKREEN STAT-SKREEN DETECTABUSE
702013 19700000-10PK 19003066-6PK 19001081-10PK 702003 702018 702004 702014 19003081-6PK 702015 19000010-6PK 19001066-10PK
76311-418PK 27.93 USD InStock
76311-418 76311-408 76311-416 76311-406 76311-414 76311-404 76311-412 76311-402 76311-410 76311-400 76311-486 76311-398
STAT-SKREEN DOA & POC Rapid Testing Controls, KOVA International
Drug Tests
Biochemical Diagnostics DETECTABUSE® STAT-SKREEN® and DOA-SKREEN liquid urine controls are designed to monitor the performance of lateral flow, rapid disposable test devices.
  • External quality DOA controls
  • Use for all lateral flow devices
  • Human urine matrix to mimic patient samples
Designed to match most testing devices on the market, with formulations updated as market changes and new testing devices become available. The STAT-SKREEN® has been the gold standard for testing since 1991 and are 510k, DEA exempt, and CE registered.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty