Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.

TI-H Multi-Frequency Ultrasonic Bath, Elma

Supplier: Elma
2390300023 2390400023 2390200023 2390100023
76326-632EA 4933.73 USD InStock
76326-632 76326-676 76326-634 76326-628
TI-H Multi-Frequency Ultrasonic Bath, Elma
Ultrasonic Cleaners
The TI-H line offers total control over the ultrasonic process. Select ultrasonic power, ultrasonic frequency, ultrasonic mode, time, and temperature. Lower frequency removes lapping and polishing media; higher frequency removes oil, grease, particles. Heavy-duty TI-H tanks are designed for 8 hours/day operation and long service life.

  • Timer with setting for continuous operation
  • Temperature adjustable 30 to 80°C
  • Ball valve drain
  • Four tank sizes from 1 to 5 gallons
  • 110 to 120 V, 50/60 Hz electrical supply
  • Powerful cleaning with power tracking for consistent performance
  • Two frequencies: 25/45 kHz for coarse or fine cleaning or mixing
  • Fully adjustable ultrasonic power

They have three ultrasonic modes for cleaning, dissolving, degassing. Extra-durable heavy-duty cleaning tank with 3-year warranty.

Certifications: CE, ISO9001, ISO13485 certified.

Ordering information: Baskets and lids are ordered separately. Choose between stainless steel lift-off lid or insulated hinged lid for noise reduction.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty