Would you like to log in again?
Ismatec Microflex Compact Single-Channel Pumps, Cole-Parmer
Supplier: Cole Parmer
Microflex®
77122-30 77122-32 77122-34
76403-104EA 1304.76 USD InStock
76403-104 76403-102 76403-100
Ismatec Microflex Compact Single-Channel Pumps, Cole-Parmer
Pumps Peristaltic Pumps
These easy-to-use complete pumps offer a wide occlusion angle, which prevents fluid backflow up to the rated pressure of the tubing. Pump fluids in either direction with easily switchable reversing motor.
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • No separate components-includes drive and pump head
  • Compact pump with single-turn speed control
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty