Would you like to log in again?
VWR® Chromatography Cart
Supplier: VWR
VWR®
91090
76318-784EA 4892.21 USD InStock
76318-784
VWR® Chromatography Cart
Carts
Made from corrosive resistant HDPE with watertight acrylic compartments, the chromatography cart was engineered specially to store and operate an HPLC machine.
  • Design for HPLC machine storage
  • Six watertight compartments for solvent storage
  • Built-in power strip
  • Holes for reagent bottle pass-through
  • Rear access opening
Engineered for chemists by chemists, the chromatography cart supports the operation of an HPLC Machine. This cart was designed with a hinged door with interior shelving, 2 drawers, and 4 locking casters. The top of the chromatography cart was built to perfect fit an HPLC machine at countertop height. This allows Auto Vial machinery to be placed directly next to the HPLC. This cart comes fully assembled, with a total lifetime replacement warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty