Would you like to log in again?
John Guest® Fittings and Adapters, Push-to-Connect , Cole-Parmer®
Supplier: Cole Parmer
34006-12 34006-06 34006-15
76377-418PK 50.38 USD InStock
76377-418 76377-422 76377-420
John Guest® Fittings and Adapters, Push-to-Connect , Cole-Parmer®
Fittings and Connectors
These single fittings offer secure connections and quick disconnection, there is no need to purchase two fitting halves.
  • For a wide variety of tubing material, including soft metals
  • Sterilize: Autoclave or ethylene oxide (ETO)
Simply push tubing past the O-ring to the internal tube stop-steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth-tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
Adapter lets to permanently modify basic fittings for customized installations. Tube-tohose stems 'S' and elbows 'T' adapt fittings for braided tubing.
Certifications: Meet NSF and FDA compliance for food-grade applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty