Would you like to log in again?
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Cole-Parmer
Supplier: Cole Parmer
77200-32 77200-62
76403-108EA 772.56 USD InStock
76403-108 76403-112
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Cole-Parmer
Pumps Peristaltic Pumps
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® High-Performance Precision pump tubing sizes
  • Automatic tubing retention
  • Fixed occlusion
These pump heads accept the four L/S® high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids.
When two Easy-Load® II pump heads are stacked rollers are offset by 45°--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
Ordering information: For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under 'Accessories'.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty