Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers and CO₂ Backup, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE40040FDRCO RDE40040LDRCO RDE50040FDRCO RDE60040LARCO RDE40040FARCO RDE40040FARCO RDE50040LARCO RDE60040LDRCO RDE50040LDRCO RDE60040FDRCO RDE60040FARCO RDE50040FARCO RDE40040LDRCO RDE40040LARCO RDE40040FDRCO RDE60040LDRCO RDE60040FARCO RDE40040LARCO RDE50040LARCO RDE50040FARCO RDE50040FDRCO RDE60040LARCO RDE50040LDRCO RDE60040FDRCO
76415-572EA 23438.4 USD InStock
76415-572 76415-584 CA76415-592 76415-540 CA76415-584 CA76415-540 CA76415-572 76415-564 76415-592 CA76415-534 CA76415-556 CA76415-578 76415-548 CA76415-600 CA76415-548 76415-528 CA76415-564 76415-578 76415-600 76415-534 76415-556 CA76415-608 CA76415-528 76415-608
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers and CO₂ Backup, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Delivery information: Package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed CO₂ backup system.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty