Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with LN₂ Back-up, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE50040FARLN RDE40040FDRLN RDE60040LDRLN RDE50040FDRLN RDE50040LARLN RDE40040LARLN RDE40040LARLN RDE50040LARLN RDE50040LDRLN RDE50040LDRLN RDE40040LDRLN RDE40040FDRLN RDE60040FARLN RDE60040FDRLN RDE40040FARLN RDE40040FARLN RDE40040LDRLN RDE60040FARLN RDE60040LDRLN RDE50040FDRLN RDE50040FARLN RDE60040LARLN RDE60040LARLN RDE60040FDRLN
76415-562EA 22821.91 USD InStock
CA76415-582 CA76415-570 76415-562 CA76415-562 76415-590 76415-570 76415-582 CA76415-590 CA76415-546 76415-526 CA76415-526 76415-538 76415-532 76415-576 76415-554 76415-598 76415-546 CA76415-532 CA76415-576 CA76415-554 CA76415-598 76415-606 CA76415-538 CA76415-606
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with LN₂ Back-up, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Delivery information: Package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed LN₂ backup system.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty