Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with Chart Recorder, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE50040FARCR RDE60040FARCR RDE50040FDRCR RDE60040LARCR RDE50040LARCR RDE50040LDRCR RDE60040LDRCR RDE60040LARCR RDE60040FDRCR RDE60040FARCR RDE50040FARCR RDE60040FDRCR RDE50040LDRCR RDE50040LARCR RDE60040LDRCR RDE50040FDRCR
76415-550EA 20744.77 USD InStock
76415-550 76415-594 76415-586 CA76415-594 76415-542 CA76415-550 76415-602 76415-558 CA76415-558 CA76415-602 CA76415-542 CA76415-586 76415-566 76415-610 CA76415-610 CA76415-566
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with Chart Recorder, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Delivery information: Package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty