Would you like to log in again?
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Supplier: Avantor Fluid Handling
78018-40 78018-42
76417-820EA 3329.76 USD InStock
76417-820 76417-818
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Pumps Peristaltic Pumps
Versatile pumps for accurate low-flow fluid delivery with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5" (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen, MasterflexLive, or analog remote control
  • Continuous run, volume dispense, and time dispense modes - batch control and interval/off time in Volume and Time modes
  • Anti-drip and speed ramping available in Volume dispense mode - ensure accuracy and minimize sample loss
  • Miniflex pump head accepts continuous tubing for clean, uninterrupted fluid paths
  • Open-head sensor
MasterflexLive® is a secure, cloud-enabled platform for control and monitoring of Masterflex pumps, get real-time control of all pump parameters - including speed, flow rate, dispense volume, and more. You can monitor and adjust critical processes running 24/7 without the need for onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via standard ethernet (RJ45) jack on rear of drive, USB connection allows for downloadable firmware updates to be loaded into the pump.
The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.
Delivery information: Includes pump drive, pump head, and power adapter with 6' (1.8 m) line cord.
Caution: Not washdown capable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty