Would you like to log in again?
Acid Surrogates Standards for US EPA Methods 625, 8270 and CLP Series Methods, VWR Chemicals BDH®
Supplier: Avantor
VWR Chemicals BDH®
BDH0743-5ML BDH0742-1ML
BDH0743-5MLEA 44.16 USD InStock
BDH0743-5ML BDH0742-1ML
Acid Surrogates Standards for US EPA Methods 625, 8270 and CLP Series Methods, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty