Would you like to log in again?
Base/Neutral Surrogate Standards for CLP Series Methods
Supplier: Avantor
VWR Chemicals BDH®
BDH0782-5ML BDH0781-1ML
BDH0781-1MLEA 128.49 USD InStock
BDH0781-1ML BDH0782-5ML
Base/Neutral Surrogate Standards for CLP Series Methods
Standards Environmental Analysis Standards
Semivolatiles standard.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty