Would you like to log in again?
Internal Standard/Surrogate, 4-Bromo-3,5-dimethylphenyl-N-methylcarbamate, for US EPA Method 531
Supplier: Avantor
VWR Chemicals BDH®
BDH0732-1ML
BDH0732-1MLEA 59.06 USD InStock
BDH0732-1ML
Internal Standard/Surrogate, 4-Bromo-3,5-dimethylphenyl-N-methylcarbamate, for US EPA Method 531
Standards Environmental Analysis Standards
Standard for pesticides, PCBs and herbicides.
The EPA is tasked with the monitoring of environmental systems as they pertain to contamination and human health. The methods issued by the EPA are created to respond to specific toxins or persistent organic pollutants (POPs) found in environmental samples such as soil, source water, drinking water, and waste. In particular, pollutants such as polychlorinated biphenyls (PCBs), which were in widespread industrial use up until their restriction, are of concern due to their stable and persistent nature in the environment. Another group of chemicals of high concern is the hundreds of commercial pesticides and herbicides in use in the world today. Pesticides, from algaecides to virucides, are used in large quantities in industrial and private agriculture. The concern over human pesticide exposure over the past few decades has led to extensive monitoring of these pesticides. It has been reported that over 98% of insecticides and 95% of herbicides affect areas other than their intended target product. It is essential that monitoring agencies have accurate standard mixes to measure the pesticide levels in the environment.
Many new pesticides are now being tested using highly sensitive LC/MS techniques, in addition to traditional GC techniques, to determine minute amounts of residue in environmental samples and food products.
US EPA Method 531 is an analytical method for the monitoring of n-methylcarbamoyloximes and n-methylcarbamates in drinking water and ground water by HPLC and a fluorescence detector
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty