Would you like to log in again?
Polynuclear Aromatic Hydrocarbon (PAH) Mix A
Supplier: Avantor
VWR Chemicals BDH®
BDH0716-1ML
BDH0716-1MLEA 140.69 USD InStock
BDH0716-1ML
Polynuclear Aromatic Hydrocarbon (PAH) Mix A
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8100 is an analytical method for the monitoring of PNA hydrocarbons in aqueous and solid samples by GC/FID.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty