Would you like to log in again?
Surrogate Standard for US EPA Method 624
Supplier: Avantor
VWR Chemicals BDH®
BDH0758-1ML
BDH0758-1MLEA 34.48 USD InStock
BDH0758-1ML
Surrogate Standard for US EPA Method 624
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty