Would you like to log in again?
MQuant® StripScan Reference Card for the Measurement of MQuant® Nitrate Test, (Cat. No.EM1.10020), EMD Millipore
Supplier: MilliporeSigma
1.03733.0001
EM1.03733.0001EA 153.56 USD InStock
EM1.03733.0001
MQuant® StripScan Reference Card for the Measurement of MQuant® Nitrate Test, (Cat. No.EM1.10020), EMD Millipore
Standards Color Reference and Spectroscopy Standards
MQuant® StripScan Reference Cards to be used in conjunction with MQuant® StripScan app. (free download). Easy-to-use analytical detection system provides reliable quantification of water, food & beverage samples for customers from research, industry, environment and diagnostics.
  • Quick, easy, and reliable
  • Superior precision – finer gradation of results
  • Convenient data analysis
The MQuant® StripScan Reference Card ensures reliable and precise results by providing an external color standard. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.
The StripScan Web application allows the user to read out your MQuant® test strips in seconds, compare with reference card and analyze, graph and share your data measurements.
This reference card allows for the digital readout of nitrate test strips 1.10020 and 1.10092 (CA only), at a concentration range between 0 mg/L and 500 mg/L nitrate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty