Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Low-Level Total Residual Chlorine Wastewater QC Sample, SPEX CertiPrep
Supplier: SPEX CertiPrep
917
SX917EA 112.18 USD InStock
SX917
Low-Level Total Residual Chlorine Wastewater QC Sample, SPEX CertiPrep
Standards Proficiency Testing Standards
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Designed for testing at low µg/L levels. Use with sensitive titrimetric or colorimetric methods.
The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and methof ruggedness.
QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty