Would you like to log in again?
DuraFrame Security Magnetic Sign Holders, Durable
Supplier: Durable
DBL4772132 DBL4772131 DBL4772130
76436-598PK 27.52 USD InStock
76436-598 76436-682 76436-490
DuraFrame Security Magnetic Sign Holders, Durable
Sign Holders
Dual-colored magnetic infoframe for all types of safety and warning information.
  • Four-sided frame works horizontally or vertically.
  • Anti-glare plastic
  • Sturdy PVC construction
  • Frame accommodates 8.5×11" insert
Dual-colored magnetic infoframe attaches to metallic surfaces such as whiteboards, metal cabinets, racking, and machinery. Sturdy cover pulls back effortlessly for speedy insert updates of safety markings, direction and warning signs such as escape and rescue routes, and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty