Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
High Performance FR Men's Dual Hazard Lightweight Pants, Navy, Lakeland Industries
Supplier: Lakeland Industries
IP8WUT13-28-30 IP8WUT13-46-34 IP8WUT13-44-28 IP8WUT13-46-30 IP8WUT13-36-28 IP8WUT13-30-28 IP8WUT13-46-32 IP8WUT13-48-28 IP8WUT13-36-36 IP8WUT13-31-34 IP8WUT13-28-28 IP8WUT13-35-32 IP8WUT13-34-36 IP8WUT13-30-32 IP8WUT13-48-32 IP8WUT13-32-34 IP8WUT13-32-32 IP8WUT13-44-32 IP8WUT13-34-32 IP8WUT13-35-30 IP8WUT13-30-34 IP8WUT13-32-28 IP8WUT13-36-32 IP8WUT13-33-30 IP8WUT13-33-32 IP8WUT13-46-28 IP8WUT13-38-32 IP8WUT13-33-36 IP8WUT13-48-34 IP8WUT13-42-32 IP8WUT13-34-34 IP8WUT13-40-32 IP8WUT13-38-28 IP8WUT13-38-34 IP8WUT13-42-34 IP8WUT13-50-30 IP8WUT13-31-30 IP8WUT13-44-30 IP8WUT13-40-34 IP8WUT13-38-30 IP8WUT13-44-34 IP8WUT13-40-30 IP8WUT13-30-30 IP8WUT13-28-34 IP8WUT13-48-30 IP8WUT13-31-32 IP8WUT13-32-36 IP8WUT13-50-34 IP8WUT13-42-28 IP8WUT13-40-28 IP8WUT13-28-32 IP8WUT13-42-30 IP8WUT13-36-30 IP8WUT13-35-34 IP8WUT13-40-36 IP8WUT13-33-34 IP8WUT13-50-28 IP8WUT13-34-28 IP8WUT13-32-30 IP8WUT13-36-34 IP8WUT13-34-30 IP8WUT13-35-36 IP8WUT13-50-32
76437-992EA 169.2 USD InStock
76437-992 76437-990 76437-906 76437-948 76437-904 76437-908 76437-942 76437-986 76437-984 76437-940 76437-902 76437-946 76437-988 76437-900 76437-944 76437-882 76437-960 76437-880 76437-916 76437-958 76437-914 76437-918 76437-952 76437-996 76437-876 76437-994 76437-950 76437-874 76437-912 76437-956 76437-954 76437-998 76437-910 76437-878 76437-890 76437-970 76437-894 76437-892 76437-928 76437-926 76437-886 76437-964 76437-920 76437-884 76437-962 76437-968 76437-924 76437-888 76437-922 76437-966 76437-982 76437-980 76437-938 76437-936 76437-974 76437-930 76437-898 76437-972 76437-896 76437-978 76437-934 76437-932 76437-976
High Performance FR Men's Dual Hazard Lightweight Pants, Navy, Lakeland Industries
Workwear
High Performance FR Lightweight Pants feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
  • Wider waistband for increased comfort
  • Articulated knees offer better mobility
  • Cell phone pocket on right leg
  • Stealth zipper pocket on right leg
  • Crotch gusset for increased comfort
  • Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The unique basketweave fabric blend provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty