Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Face Shield
Supplier: United Scientific Supplies
FSMPE-PE
470329-612EA 3.95 USD InStock
470329-612
Face Shield
Face Shields
Face shield provides reliable splash protection in a variety of applications and environments.
  • Covers the entire face
  • Non sterile
  • Anti-fog coating
  • Manufactured in an FDA registered facility
Safe, comfortable, and easy to use. This facemask can be worn with or without goggles or glasses. It offers reliable protection and features a soft elastic band, padded headband, and clear PET shield. Each shield is individually wrapped, and can either be disposed of or sanitized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty