Would you like to log in again?
SP Wilmad-LabGlass NMR Tip-off Manifolds, SP Industries
Supplier: SP Industries
552-3 552-4 552-5
76459-874EA 163.53 USD InStock
76459-874 76459-876 76459-878
SP Wilmad-LabGlass NMR Tip-off Manifolds, SP Industries
Manifolds
Allows for easy connection between NMR and EPR tubes to a vacuum line.
  • Highly resistant to chemicals
  • Easy operation
Rotating the valve opens and closes the tube to the vacuum line.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty