Would you like to log in again?
NM522 Competent Cells, Agilent Technologies
Supplier: Agilent Technologies
200233 200233
99900-336EA 212.14 USD InStock
CA99900-336 99900-336
NM522 Competent Cells, Agilent Technologies
Competent Cells
NM522 competent cells are recombination-proficient and EcoK restriction-deficient.
  • Recombination-proficient and EcoK restriction-deficient
  • Blue-white color screening possible for recombinant plasmids
  • Transformation efficiency >1×10⁸ transformants/µg DNA
NM6522 competent cells are a classic E. coli strain for economical cloning when you don’t need supercompetent efficiencies. NM522 is a recombination-proficient and EcoK restriction deficient host strain. Intrinsic antibiotic resistance aids cell selection after a complete and successful transformation.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty