Would you like to log in again?
Mopec Bone Dust Vacuum, Mortech®
Supplier: Mortech
BD080
76498-346EA 6718.67 USD InStock
76498-346
Mopec Bone Dust Vacuum, Mortech®
Vacuum Cleaners
HEPA filtration minimizes exposure to particulate matter. The bone dust vacuum collector attaches directly to the autopsy saw for safety and efficiency.
  • One-year warranty
  • 115 V, 60 Hz with US plug
  • Reduced noise level
  • HE-rated filter is powerful enough for even the smallest particles
  • Disposable filter cartridge is easy to replace
  • 10′ power cord
HEPA vacuum includes HEPA filter, vacuum hose (102"), and 'Y' bracket. The vacuum is designed specifically for use with Stryker 810 Autopsy Saw, Mopec 810 Autopsy Saw, and Mopec 1000 Autopsy Saw. Not for use with Mopec 5000 Autopsy Saw.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty