Would you like to log in again?
VWR® Standard Manual Defrost Laboratory Freezers with Natural Refrigerants
Supplier: VWR
VWR®
HCMFS-20
75836-238EA 4034.49 USD InStock
75836-238
VWR® Standard Manual Defrost Laboratory Freezers with Natural Refrigerants
Freezers Laboratory / General Purpose Freezers
These efficient and economical freezers provides quality cold storage in laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant (R290)
These manual defrost freezers offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness. These freezers are equipped with keyed door locks, interior shelving, probe access, and microprocessor temperature controllers.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. The freezers run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certifications: UL or ETL Listed.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty