Would you like to log in again?
VWR® Standard Hazardous Location (Explosion Proof) Freezers with Natural Refrigerants
Supplier: Avantor
VWR®
HCEFV-17 HCEFV-14
76499-008EA 5750.08 USD InStock
76499-008 76499-006
VWR® Standard Hazardous Location (Explosion Proof) Freezers with Natural Refrigerants
Freezers Flammable Material Freezers
These freezers are engineered for cold-storage use in environments where volatile conditions could potentially exist.
  • Microprocessor temperature controller
  • High density urethane foam cabinet and door insulation
  • Interior shelving
  • Keyed door lock
  • Natural, hydrocarbon refrigerant (R290)
These freezers are equipped with keyed door locks, interior shelving, and microprocessor temperature controllers. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certifications: ETL Listed. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.
Caution: Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see VWR® Flammable Storage Units.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty