Would you like to log in again?
VWR® Standard Flammable Storage Refrigerators with Natural Refrigerants
Supplier: Avantor
VWR®
HCFRV-14
76498-992EA 4517.34 USD InStock
76498-992
VWR® Standard Flammable Storage Refrigerators with Natural Refrigerants
Refrigerators
These refrigerators are designed to store and protect flammable substances in laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant (R290)
These refrigerators utilize ATEX and non-sparking components for the safe storage of flammable substances. All units feature a series of temperature alarms to keep stored products at peak effectiveness. These refrigerators are equipped with keyed door locks, interior shelving, probe access, and microprocessor temperature controllers.
All units include a two year parts and labor warranty. The refrigerators run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certifications: ETL Listed.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty