Would you like to log in again?
HEPA Filter, Disposable, for TRIO.BAS™ HEPA Filter Equipped Air Sampler Units by Orum International, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS173
76470-196PK 450.71 USD InStock
76470-196
HEPA Filter, Disposable, for TRIO.BAS™ HEPA Filter Equipped Air Sampler Units by Orum International, Hardy Diagnostics
Samplers Air Samplers
The TRIO.BAS™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.
An attached HEPA filtration system filters the exhausted air. High efficiency filter for airborne bacteria to replace used filters in TRIO.BAS™ models 76420-906 and 76420-908.
Note: The air sampler instrument in the images is not included.
Packaging: 10 filters per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty