Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acridine orange (chlorid salt), hydrate ∼80% (dye content), pure

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acridine orange hydrochlorideBasic Orange 14N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochlorideC.I. 460053,6-Bis(dimethylamino)acridine hydrochloride3,6-AcridinediamineAO

42334-0010
TS42334-0010EA 81.91 USD InStock
TS42334-0010
Acridine orange (chlorid salt), hydrate ∼80% (dye content), pure
Acridine orange (chlorid salt)


Formula: C₁₇H₁₉N₃·HCl·xH₂O
MW: 301.81 g/mol (anhy)
Melting Pt: 284…287 °C
Storage Temperature: Ambient
MDL Number: MFCD00012660
CAS Number: 65-61-2
EINECS: 200-614-0
Order Now
Specification Test Results
Appearance Orange to brown Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
UV spectrophotometry ≥75 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty