Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Citalopram oxalate

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate

46234-0010
TS46234-0010EA 212 USD InStock
TS46234-0010
(S)-Citalopram oxalate
(S)-Citalopram oxalate


Formula: C₂₀H₂₁FN₂O
MW: 414.43 g/mol
Melting Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06407826
CAS Number: 219861-08-2
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Solubility (2 % in DMSO) Clear colorless solution
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty