Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nebivolol hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

46249-2500
TS46249-2500EA 169 USD InStock
TS46249-2500
Nebivolol hydrochloride
Nebivolol hydrochloride


Formula: C₂₂H₂₅F₂NO₄
Melting Pt: 226 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07357273
CAS Number: 152520-56-4
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥97.5 % (mixture of isomers)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty