Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,6-Hexanedithiol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,6-Dimercaptohexane

12067-0250 12067-0050
TS12067-0050EA 50 USD InStock
TS12067-0050 TS12067-0250
1,6-Hexanedithiol 97%
1,6-Hexanedithiol


Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 237 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.99 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004910
CAS Number: 1191-43-1
EINECS: 214-735-1
UN: 3334
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5150 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +16° to +18° (20°C, 589 nm) (c=1,H2O)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty