Would you like to log in again?
Two-Way, Three-Way, and Sample Diaphragm Valves
Supplier: Parker
MV-10-1044-02 QV2-186-M MV-10-1344-02 MV-10-1344-00 MV-11-005 201-48
MFLX30502-07EA 579 USD InStock
MFLX30502-07 MFLX31101-03 MFLX98402-08 MFLX98403-08 MFLX31101-05 MFLX31101-07
Two-Way, Three-Way, and Sample Diaphragm Valves
Valves Diaphragm Valves
Easily control flow with diaphragm valve.
  • Constructed of chemically inert PFA for a metal-free degsign
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty