Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Toluidine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Aminotoluene2-Methylaniline2-Toluidine

13908-5000 13908-0100 13908-0025
TS13908-0100EA 474 USD InStock
TS13908-0100 TS13908-5000 TS13908-0025
o-Toluidine 99%
o-Toluidine


Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 199…200 °C (760 mmHg)
Melting Pt: –23 °C
Density: 1 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS Number: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5710 to 1.5730 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty