Would you like to log in again?
United Scientific™ Porcelain Desiccator Plates, Small Holes, United Scientific Supplies
Supplier: United Scientific Supplies
JSD140 JSD190 JSD230
470340-204EA 36.35 USD InStock
470340-204 470339-954 470340-060
United Scientific™ Porcelain Desiccator Plates, Small Holes, United Scientific Supplies
Desiccators Desiccators, Plates
Porcelain desiccator plates with small holes.
  • Autoclavable
  • Center hole is 25 mm in diameter, surrounded by various 5 mm holes
  • Glazed on top surface; no glaze softening occurs up to 1000 °C
Porcelain desiccator plate with 140 mm diameter. Features numerous perforations: center hole is 25 mm in diameter, and the remaining are each 5 mm in diameter. Plate is glazed on top surface and is resistant to acids and alkalies, excluding hydrofluoric acid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty